Learn More
Medchemexpress LLC E7766 diammonium salt | 2242635-03-4 | 99.48% | 780.66 | 1 MG
SDP
Catalog No. 5000365455
Encompass_Preferred
E7766 diammonium salt is a macrocycle-bridged STING agonist with a Kd of 40 nM. It demonstrates potent pan-genotypic and antitumor activities.
  • Macrocycle-bridged STING agonist
  • Kd of 40 nM
  • Shows potent pan-genotypic and antitumor activities
  • Inhibits four human STING variants (human wild-type, HAQ, AQ, and REF STING proteins) with EC50 values of 1 μM, 2.2 μM, 1.2 μM, and 4.9 μM, respectively
  • Demonstrates potent antitumor activity with a long-lasting immune memory response in a mouse liver metastatic tumor model
  • For research use only
  • Appearance: Solid
  • Color: White to off-white
Catalog No. 50-003-65455 Supplier Medchemexpress LLC Supplier No. 1MGHY111999A
E7766 diammonium salt is a macrocycle-bridged STING agonist with a Kd of 40 nM. It demonstrates potent pan-genotypic and antitumor activities.
  • Macrocycle-bridged STING agonist
  • Kd of 40 nM
  • Shows potent pan-genotypic and antitumor activities
  • Inhibits four human STING variants (human wild-type, HAQ, AQ, and REF STING proteins) with EC50 values of 1 μM, 2.2 μM, 1.2 μM, and 4.9 μM, respectively
  • Demonstrates potent antitumor activity with a long-lasting immune memory response in a mouse liver metastatic tumor model
  • For research use only
  • Appearance: Solid
  • Color: White to off-white