Learn More
Medchemexpress LLC E7766 diammonium salt | 2242635-03-4 | 99.5% | 780.66 | 10 MG
SDP
Catalog No. 5000437645
Encompass_Preferred
E7766 diammonium salt is a macrocycle-bridged STING agonist with a Kd of 40 nM. It demonstrates potent pan-genotypic and antitumor activities. For research use only, not for sale to patients.
  • Macrocycle-bridged STING agonist
  • Kd of 40 nM
  • Potent pan-genotypic activity
  • Potent antitumor activities
  • Inhibits four human STING variants (human wild-type, HAQ, AQ, and REF STING proteins) with EC50 values of 1 μM, 2.2 μM, 1.2 μM, and 4.9 μM, respectively
  • Shows potent antitumor activity with long-lasting immune memory response in a mouse liver metastatic tumor model
Catalog No. 50-004-37645 Supplier Medchemexpress LLC Supplier No. HY111999A10MG
E7766 diammonium salt is a macrocycle-bridged STING agonist with a Kd of 40 nM. It demonstrates potent pan-genotypic and antitumor activities. For research use only, not for sale to patients.
  • Macrocycle-bridged STING agonist
  • Kd of 40 nM
  • Potent pan-genotypic activity
  • Potent antitumor activities
  • Inhibits four human STING variants (human wild-type, HAQ, AQ, and REF STING proteins) with EC50 values of 1 μM, 2.2 μM, 1.2 μM, and 4.9 μM, respectively
  • Shows potent antitumor activity with long-lasting immune memory response in a mouse liver metastatic tumor model