Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC E7766 diammonium salt | 2242635-03-4 | 99.5% | 780.66 | C24H32F2N12O8P2S2 | 5 MG
SDP

Catalog No. 5000263698
Encompass_Preferred

E7766 diammonium salt is a macrocycle-bridged stimulator of interferon genes (STING) agonist that binds STING with high affinity (Kd ≈ 40 nM) and has shown potent pan-genotypic activation and antitumor efficacy in preclinical studies. The compound is supplied as the diammonium salt for research applications and is provided at high reported purity with recommended frozen storage to preserve stability.

  • Macrocycle-bridged STING agonist with high affinity (Kd ≈ 40 nM).
  • Demonstrated pan-genotypic STING activation and antitumor activity in preclinical models.
  • Supplied as a diammonium salt with high purity (≈99.5%).
  • Molecular weight 780.66 g·mol-1; formula C24H32F2N12O8P2S2.
  • Recommended storage at -20°C to maintain stability for research use.

Catalog No. 50-002-63698 Supplier Medchemexpress LLC Supplier No. HY111999A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

E7766 diammonium salt is a macrocycle-bridged stimulator of interferon genes (STING) agonist that binds STING with high affinity (Kd ≈ 40 nM) and has shown potent pan-genotypic activation and antitumor efficacy in preclinical studies. The compound is supplied as the diammonium salt for research applications and is provided at high reported purity with recommended frozen storage to preserve stability.

  • Macrocycle-bridged STING agonist with high affinity (Kd ≈ 40 nM).
  • Demonstrated pan-genotypic STING activation and antitumor activity in preclinical models.
  • Supplied as a diammonium salt with high purity (≈99.5%).
  • Molecular weight 780.66 g·mol-1; formula C24H32F2N12O8P2S2.
  • Recommended storage at -20°C to maintain stability for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.