Learn More
Grainger Vikan Squeegee Refill, Hygienic, TPE Rubber, 23.6 in., Black, 1 ea.
Catalog No. 502787010
Encompass_Preferred
This hygienic replacement squeegee blade is designed for use with fixed and revolving neck squeegees (Series 7714x and 7724x) to maintain optimal cleanliness in food manufacturing, food service, and other sensitive environments. Featuring a flexible double-blade design, this cassette effectively removes excess water and debris from tiled or uneven anti-slip surfaces.
  • Double blade design balances hygiene and efficiency for superior debris removal
  • Flexible construction enables the blade to follow surface contours
  • Easily removable cassette simplifies cleaning and reduces maintenance costs
  • Durable, chemical-resistant material withstands daily use with cleaning agents
  • Color-coded to support hygienic zoning plans and cross-contamination reduction
  • FDA-compliant and meets international food contact safety regulations
Catalog No. 50-278-7010 Supplier Grainger Supplier No. 18G948
This hygienic replacement squeegee blade is designed for use with fixed and revolving neck squeegees (Series 7714x and 7724x) to maintain optimal cleanliness in food manufacturing, food service, and other sensitive environments. Featuring a flexible double-blade design, this cassette effectively removes excess water and debris from tiled or uneven anti-slip surfaces.
  • Double blade design balances hygiene and efficiency for superior debris removal
  • Flexible construction enables the blade to follow surface contours
  • Easily removable cassette simplifies cleaning and reduces maintenance costs
  • Durable, chemical-resistant material withstands daily use with cleaning agents
  • Color-coded to support hygienic zoning plans and cross-contamination reduction
  • FDA-compliant and meets international food contact safety regulations