Learn More
Medchemexpress LLC HY-14571 5mg , E7820 CAS:289483-69-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY145715MG
Encompass_Preferred
HY-14571 5mg E7820 CAS: 289483-69-8 E7820 (ER68203-00), an orally active aromatic sulfonamide derivative, is a unique angiogenesis inhibitor suppressing an expression of integrin alpha2 subunit on endothelium. E7820 inhibits rat aorta angiogenesis with an IC50 of 0.11 μg/ml. E7820 modulates α-1, α-2, α-3, and α-5 integrin mRNA expression. Antiangiogenic and antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9520