Learn More
Abcam E7820, 5MG
Catalog No. 502680458
Encompass_Preferred
MW 336.4 Da, Purity >=98%. A small molecule sulfonamide derivative with potential antiangiogenic and antitumor activities. E7820 inhibits angiogenesis by suppressing integrin alpha 2, a cell adhesion molecule expressed on endothelial cells. Inhibition of integrin alpha 2 leads to an inhibition of cell-cell interactions, endothelial cell-matrix interactions, vascular endothelial cell proliferation and angiogenesis.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-268-0458 Supplier Abcam Supplier No. AB2871065MG
MW 336.4 Da, Purity >=98%. A small molecule sulfonamide derivative with potential antiangiogenic and antitumor activities. E7820 inhibits angiogenesis by suppressing integrin alpha 2, a cell adhesion molecule expressed on endothelial cells. Inhibition of integrin alpha 2 leads to an inhibition of cell-cell interactions, endothelial cell-matrix interactions, vascular endothelial cell proliferation and angiogenesis.
The product is subject to the following: Abcam Restricted Use Statement