Learn More
Medchemexpress LLC HY-10045 5mg Medchemexpress, AEE788 CAS:497839-62-0 Purity:>98%
SDP
Catalog No. 501931838
Encompass_Preferred
Medchemexpress, HY-10045 5mg AEE788 CAS:497839-62-0 AEE788 is an inhibitor of the EGFR and ErbB2 with IC 50 values of 2 and 6 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1838 Supplier Medchemexpress LLC Supplier No. HY100455MG
Medchemexpress, HY-10045 5mg AEE788 CAS:497839-62-0 AEE788 is an inhibitor of the EGFR and ErbB2 with IC 50 values of 2 and 6 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.