Learn More
LW Scientific E8 Touch Screen 8-place Centrifuge
Catalog No. 22500135 Shop All LW Scientific Products
Change view
Click to view available options
Capacity:
8-place
Catalog No. Capacity
22-500-135 8-place
1 options
This item is not returnable. View return policy
Catalog No. 22-500-135 Supplier LW Scientific Supplier No. E8CU8AT15T3
Add to Cart
Add to Cart
This item is not returnable. View return policy
E8 Touch Screen 8-place Centrifuge holds 3 to 15 mL test tubes.
  • This model is loaded with features and improvements, such as higher speeds for faster and cleaner separations, quieter operation for peaceful work environments, lower chamber temperatures for cooler test-tube samples, shorter height to fit under any cabinet, and auto-calibrating speed function for precision results.
  • It even has an auto-brake to stop the rotor in less than 30 seconds to save time in the busy labs.
  • The E8 family also includes a digital, touchscreen model, with no knobs and precise time and speed programs for CLIA compliance and the highest accuracy at an affordable price.
  • The E8 Fixed is designed for blood separations at a fixed speed of 3500 rpm only (1534 g-force).
  • The E8 Variable and E8 Touch will separate blood at 3500 rpm and can be slowed to separate other fluids at lower g-force, such as urine specimens as outlined by C.L.I.A. regulations.
Specifications
Capacity 8-place
Max. RCF 1,534 x g
Max. Speed 3,500 rpm
Refrigerated No
Memory Programmable
Voltage 110/220 V