Learn More
LW Scientific E8 Touch Screen Combination 8-place Centrifuge
Catalog No. 22500137 Shop All LW Scientific Products
Change view
Click to view available options
Capacity:
8-place
Catalog No. Capacity
22-500-137 8-place
1 options
This item is not returnable. View return policy
Catalog No. 22-500-137 Supplier LW Scientific Supplier No. E8CU8ATCRT3
Add to Cart
Add to Cart
This item is not returnable. View return policy
E8 Touch Screen Combination 8-place Centrifuge is ideal for spinning test tubes, micro tubes and microhematocrit tubes.
  • Spin test tubes, micro tubes and microhematocrit tubes in one centrifuge with no rotor changes!
  • The E8 Combination Centrifuge is specifically designed to run 75 mm microhematocrit tubes and 1 mL to 15 mL micro tubes and test tubes.
  • This centrifuge features a quiet operation for a more peaceful laboratory environment.
  • It also features a shorter height to fit under any cabinet, and auto-calibrating speed function for precision results.
  • To run microhematocrit tubes, simply insert your 75 mm capillary tubes into the carrier, place carriers in opposing tubes sleeves for a balanced load, and spin at 3,400 rpm (or higher) for 6 (± 1) minutes for clear separation.
Specifications
Capacity 8-place
Max. RCF 1,534 x g
Max. Speed 3,500 rpm
Refrigerated No
Memory Programmable
Voltage 110/220 V