Learn More
LW Scientific E8 Touch Screen Portable 8-place Centrifuge
Catalog No. 22500136 Shop All LW Scientific Products
Change view
Click to view available options
Capacity:
8-place
Catalog No. Capacity
22-500-136 8-place
1 options
This item is not returnable. View return policy
Catalog No. 22-500-136 Supplier LW Scientific Supplier No. E8CU8AT15TP
Add to Cart
Add to Cart
This item is not returnable. View return policy
E8 Touch Screen Portable 8-place Centrifuge is ideal for spinning on the go.
  • The portable E8 Portafuge is designed for spinning on the go.
  • This model is loaded with features and improvements, such as higher speeds for faster and cleaner separations, quieter operation, lower chamber temperatures for cooler test-tube samples, and auto-calibrating function for precision speeds.
  • It even has an auto-brake to stop the rotor in less than 20 seconds to save time.
  • Simply place the centrifuge into the included car seat, strap the car seat into your vehicle, and insert the power outlet cord into your car’s 12v power outlet.
  • Optional wall adapter not included.
Specifications
Capacity 8-place
Max. RCF 1,534 x g
Max. Speed 3,500 rpm
Refrigerated No
Memory Programmable
Voltage 110/220 V