Learn More
Enzo Life Sciences Indirubin-3'-(2,3-dihydroxypropyl)oximether (1mg)
SDP
Catalog No. 502008549
Encompass_Preferred
Inhibitor of c-Src (IC50=0.43µM) blocking downstream STAT3 signalling. Induces apoptosis in human cancer cells. Alternative name: E804. Purity: ≥96% (TLC). Appearance: Dark red solid. Solubility: Soluble in DMSO (1M). Long Term Storage: -20°C.
Catalog No. 50-200-8549 Supplier Enzo Life Sciences Supplier No. ALX270425M001
Inhibitor of c-Src (IC50=0.43µM) blocking downstream STAT3 signalling. Induces apoptosis in human cancer cells. Alternative name: E804. Purity: ≥96% (TLC). Appearance: Dark red solid. Solubility: Soluble in DMSO (1M). Long Term Storage: -20°C.