Learn More
Cayman Chemical E8Methyl6nonoIc AcId 100mg
SDP
Catalog No. 502966288
Encompass_Preferred
A potential thermal degradant of capsaicin with possible mutagenic effects
Catalog No. 50-296-6288 Supplier Cayman Chemical Supplier No. 16609100 MG
A potential thermal degradant of capsaicin with possible mutagenic effects