Learn More
Cayman Chemical E8Methyl6nonoIc AcId 500mg
SDP
Catalog No. 502966112
Encompass_Preferred
A potential thermal degradant of capsaicin with possible mutagenic effects
Catalog No. 50-296-6112 Supplier Cayman Chemical Supplier No. 16609500 MG
A potential thermal degradant of capsaicin with possible mutagenic effects