Learn More
Volu Sol Papanicolaou Stain, EA-65, 32 oz / 1 L, Volu-Sol™
SDP
Supplier:  Volu Sol VSE032
Encompass_Preferred
Papanicolaou stain used for critical examination of nuclei and cytoplasmic components of exfoliative cells
Catalog No. 50-420-840