Learn More
StatLab™ EA-65 Cytology Stain
GSA_VASDP
Catalog No. STLSL302
Change view
Click to view available options
Quantity:
4 x 500 mL
Catalog No. Quantity
STLSL302 4 x 500 mL
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STLSL302 Supplier StatLab™ Supplier No. SL302
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Alcohol-based, for routine cytoplasmic staining.
  • Cytological counterstain used for non-gynecological cytodiagnosis in Papanicolaou staining technique
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Quantity 4 x 500 mL
Product Type Cytology Stain