Learn More
Electron Microscopy Sciences EA Counterstain, Gill's Modified 500 ML
SDP
Catalog No. 5025692
Encompass_Preferred
EA Counterstain, Gill's Modified solution is a prepared, ready-to-use, high quality staining solution that saves time and effort and produces consistent uniform results. For use with Papanicolaou stain; a general tissue stain.
Catalog No. 50-256-92 Supplier Electron Microscopy Sciences Supplier No. 2605005
EA Counterstain, Gill's Modified solution is a prepared, ready-to-use, high quality staining solution that saves time and effort and produces consistent uniform results. For use with Papanicolaou stain; a general tissue stain.