Learn More
Electron Microscopy Sciences EA Counterstain, Gill's Modified 1 LT
SDP
Catalog No. 5025691
Encompass_Preferred
EA Counterstain, Gill's Modified solution is a prepared, ready-to-use, high quality staining solution that saves time and effort and produces consistent uniform results. For use with Papanicolaou stain; a general tissue stain.
Catalog No. 50-256-91 Supplier Electron Microscopy Sciences Supplier No. 2605006
EA Counterstain, Gill's Modified solution is a prepared, ready-to-use, high quality staining solution that saves time and effort and produces consistent uniform results. For use with Papanicolaou stain; a general tissue stain.