Learn More
Medchemexpress LLC EAAT2 activator 1 | 892415-28-0 | 98.9% | 331.80 | 50 MG
SDP
Catalog No. 5000351832
Encompass_Preferred
EAAT2 activator 1 is a potent activator of excitatory amino acid transporter 2 (EAAT2). EAAT2 is the primary glutamate transporter responsible for removing glutamate from synapses. This activator increases EAAT2 protein levels in a dose-dependent manner.
  • Potent activator of EAAT2
  • Removes glutamate from synapses
  • Increases protein levels dose-dependently
  • Solid appearance
  • Light brown to brown color
Catalog No. 50-003-51832 Supplier Medchemexpress LLC Supplier No. HY13969250MG
EAAT2 activator 1 is a potent activator of excitatory amino acid transporter 2 (EAAT2). EAAT2 is the primary glutamate transporter responsible for removing glutamate from synapses. This activator increases EAAT2 protein levels in a dose-dependent manner.
  • Potent activator of EAAT2
  • Removes glutamate from synapses
  • Increases protein levels dose-dependently
  • Solid appearance
  • Light brown to brown color