Learn More
ABclonal Technology EAAT2 Rabbit mAb
SDP
Catalog No. 5000010621
Encompass_Preferred
This gene encodes a member of a family of solute transporter proteins The membrane-bound protein is the principal transporter that clears the excitatory neurotransmitter glutamate from the extracellular space at synapses in the central nervous system Glutamate clearance is necessary for proper synaptic activation and to prevent neuronal damage from excessive activation of glutamate receptors Improper regulation of this gene is thought to be associated with several neurological disorders Alternatively spliced transcript variants of this gene have been identified
Catalog No. 50-000-10621 Supplier ABclonal Technology Supplier No. A25213100UL
This gene encodes a member of a family of solute transporter proteins The membrane-bound protein is the principal transporter that clears the excitatory neurotransmitter glutamate from the extracellular space at synapses in the central nervous system Glutamate clearance is necessary for proper synaptic activation and to prevent neuronal damage from excessive activation of glutamate receptors Improper regulation of this gene is thought to be associated with several neurological disorders Alternatively spliced transcript variants of this gene have been identified