Learn More
Adam Equipment EAB 224e - Equinox Analytical and Semi-Micro Balances - Capacity: 220g, Readability: 0.0001g
Catalog No. NC3964790
Encompass_Preferred
Equinox analytical balances deliver remarkable precision to labs that perform highly detailed testing. The equinox is ideal for research and quality assurance facilities; science education; precision counting; and production and manufacturing applications needing exceptionally precise results. Eight analytical models offer capacities from 120G to 510G and 0.1mg readability. A color touch screen display provides intuitive operation, with icon-driven menus and readily accessible applications and functions. The five-inch screen is generously sized, offering 800 x 480 pixel resolution for excellent visibility.
Catalog No. NC3964790 Supplier Adam Equipment Supplier No. EAB 224E
Equinox analytical balances deliver remarkable precision to labs that perform highly detailed testing. The equinox is ideal for research and quality assurance facilities; science education; precision counting; and production and manufacturing applications needing exceptionally precise results. Eight analytical models offer capacities from 120G to 510G and 0.1mg readability. A color touch screen display provides intuitive operation, with icon-driven menus and readily accessible applications and functions. The five-inch screen is generously sized, offering 800 x 480 pixel resolution for excellent visibility.