Learn More
Medchemexpress LLC HY-129111 5mg Medchemexpress, EACC CAS:864941-31-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1291115MG
Encompass_Preferred
Medchemexpress, HY-129111 5mg EACC CAS:864941-31-1 EACC is a reversible autophagy inhibitor, which can block autophagic flux. EACC selectively inhibits the translocation of autophagosome-specific SNARE Stx17 thereby blocking autophagosome-lysosome fusion. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9797