Learn More
Med Vet International Sterile Water For Injection, 10mL Vial
Catalog No. 502828804
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Nonpyrogenic, distilled water in a single dose container for intravenous administration after addition of a suitable solute. It may also be used as a dispensing container for diluent use. No antimicrobial or other substance has been added. The pH is 5.5 (5.0 to 7.0).
Catalog No. 50-282-8804 Supplier Med Vet International Supplier No. RXWATER10
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Nonpyrogenic, distilled water in a single dose container for intravenous administration after addition of a suitable solute. It may also be used as a dispensing container for diluent use. No antimicrobial or other substance has been added. The pH is 5.5 (5.0 to 7.0).