Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Eact | 461000-66-8 | 98.9% | 428.50 g/mol | C22H24N2O5S | 50 MG
SDP

Catalog No. 5000244951
Encompass_Preferred

Eact is a small-molecule research compound that selectively activates the TMEM16A chloride channel and directly activates TRPV1 channels in sensory nociceptors, producing itch, acute nociception, and thermal hypersensitivity. It is supplied as a high-purity powder for in vitro and in vivo pharmacology studies.

  • Selective TMEM16A activator and TRPV1 activator.
  • High purity (98.86%).
  • Molecular weight 428.50 g/mol; formula C22H24N2O5S.
  • Soluble in DMSO (50 mg/mL); in vivo formulation example 10% DMSO/90% corn oil.
  • Recommended storage: powder at -20°C or 4°C; solutions at -80°C or -20°C.
  • Suitable for in vitro and in vivo pharmacology studies.
  • Available as a 50 mg pack.

Catalog No. 50-002-44951 Supplier Medchemexpress LLC Supplier No. HY10336850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Eact is a small-molecule research compound that selectively activates the TMEM16A chloride channel and directly activates TRPV1 channels in sensory nociceptors, producing itch, acute nociception, and thermal hypersensitivity. It is supplied as a high-purity powder for in vitro and in vivo pharmacology studies.

  • Selective TMEM16A activator and TRPV1 activator.
  • High purity (98.86%).
  • Molecular weight 428.50 g/mol; formula C22H24N2O5S.
  • Soluble in DMSO (50 mg/mL); in vivo formulation example 10% DMSO/90% corn oil.
  • Recommended storage: powder at -20°C or 4°C; solutions at -80°C or -20°C.
  • Suitable for in vitro and in vivo pharmacology studies.
  • Available as a 50 mg pack.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.