Learn More
Medchemexpress LLC Eact | 461000-66-8 | MFCD03115431 | 98.9% | 428.50 g·mol⁻¹ | C22H24N2O5S | 100 MG
SDP
Catalog No. 5000244927
Encompass_Preferred
Eact is a small-molecule activator of the calcium-activated chloride channel TMEM16A (ANO1) that also directly activates TRPV1 channels in sensory nociceptors, producing itch, acute nociception, and thermal hypersensitivity. It is provided as a research-grade compound for use in pharmacological studies and mechanistic investigations.
  • Selective activator of TMEM16A (ANO1).
  • Directly activates TRPV1 in sensory nociceptors.
  • Used to model itch, nociception, and thermal hypersensitivity in research.
  • High purity (~98.9%) appropriate for in vitro and in vivo experiments.
  • Soluble in DMSO; available in multiple pack sizes including 100 mg.
Catalog No. 50-002-44927 Supplier Medchemexpress LLC Supplier No. HY103368100MG
Eact is a small-molecule activator of the calcium-activated chloride channel TMEM16A (ANO1) that also directly activates TRPV1 channels in sensory nociceptors, producing itch, acute nociception, and thermal hypersensitivity. It is provided as a research-grade compound for use in pharmacological studies and mechanistic investigations.
  • Selective activator of TMEM16A (ANO1).
  • Directly activates TRPV1 in sensory nociceptors.
  • Used to model itch, nociception, and thermal hypersensitivity in research.
  • High purity (~98.9%) appropriate for in vitro and in vivo experiments.
  • Soluble in DMSO; available in multiple pack sizes including 100 mg.