Learn More
Alpco Ltd EAEC PCR Kit
SDP
Catalog No. NC2284601
Encompass
The EAEC PCR Kit is a multiplex real-time PCR assay for the direct, qualitative detection of enteroaggregative E. coli (EAEC) in human stool samples. Research Use Only. Not for Use in Diagnostic Procedures. This kit is sufficient for 100 tests.
This item is not returnable. View return policy
Catalog No. NC2284601 Supplier Alpco Ltd Supplier No. 86EECHU100
The EAEC PCR Kit is a multiplex real-time PCR assay for the direct, qualitative detection of enteroaggregative E. coli (EAEC) in human stool samples. Research Use Only. Not for Use in Diagnostic Procedures. This kit is sufficient for 100 tests.