Learn More
New England Biolabs, Inc. EaeI – 200 units
SDP
Catalog No. 50812154
Encompass
A restriction endonuclease that recognizes the sequence Y^GGCC_R.
Catalog No. 50-812-154 Supplier New England Biolabs, Inc. Supplier No. R0508S
A restriction endonuclease that recognizes the sequence Y^GGCC_R.