Learn More
Aviva Systems Biology Corp EAF2 Antibody - C-terminal region (ARP64761_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64761_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EAF2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human EAF2. Alternate names: BM040, TRAITS, U19ApplicationWB. Species reactivity: Human
Catalog No. 50-288-361