Learn More
MSC Eagle Column Protector, 8 in Square, 42 in, 1 ea
Catalog No. 5000803614
Encompass
Column protectors provide affordable protection for columns and beams in warehouses and factories. They absorb impact from tow motors, minimizing damage to columns, fork trucks, and personnel. These protectors are blow-molded from 100 percent high-density polyethylene (HDPE), offering resistance to most chemicals.
  • Absorbs forklift and tow-motor impacts
  • Minimizes column, fork truck, and personnel damage
  • Constructed from heavy duty, blow-molded 100% high-density polyethylene (HDPE)
  • Resistant to most chemicals
  • Key-lock design prevents slippage and aids installation alignment
  • No tools required for installation
  • Two halves attach with easy-to-install black straps (included)
  • Lightweight for ease of transport
  • Stackable for easy storage
Catalog No. 50-008-03614 Supplier MSC Supplier No. 01245778
Column protectors provide affordable protection for columns and beams in warehouses and factories. They absorb impact from tow motors, minimizing damage to columns, fork trucks, and personnel. These protectors are blow-molded from 100 percent high-density polyethylene (HDPE), offering resistance to most chemicals.
  • Absorbs forklift and tow-motor impacts
  • Minimizes column, fork truck, and personnel damage
  • Constructed from heavy duty, blow-molded 100% high-density polyethylene (HDPE)
  • Resistant to most chemicals
  • Key-lock design prevents slippage and aids installation alignment
  • No tools required for installation
  • Two halves attach with easy-to-install black straps (included)
  • Lightweight for ease of transport
  • Stackable for easy storage