Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eagle™ Drum-Containment Pallets and Platforms
GREENER_CHOICE

Catalog No. 19045912 Shop All Eagle Manufacturing Products
Change view
Click to view available options
Type:
2 Drum Pallet, with Drain
4 Drum Pallet, with Drain
4 Drum Pallet, without Drain
Poly Pallet Ramp
Height (English):
6.5 in.
8 in.
13.75 in.
Width (English):
32 in.
51 in.
51.5 in.
103.5 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Height (English) Width (English)
19-045-912 2 Drum Pallet, with Drain 13.75 in. 51 in.
19-033-516 4 Drum Pallet, without Drain 6.5 in. 103.5 in.
18-999-4523 Poly Pallet Ramp 8 in. 32 in.
18-999-4470 4 Drum Pallet, with Drain 8 in. 51.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-045-912 Supplier Eagle™ Supplier No. 1620
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Grating removes easily for cleaning

Pallets and platforms contain spills and overflows of hazardous materials, keeping floors contamination-free.

  • Solid polyethylene construction provides excellent chemical resistance
  • Consist of translucent tub in safety yellow, and black grating with 1.75 in. (4.4 cm) square openings
  • Pallet units only are forkliftable
  • Poly ramp can be used with 4-drum inline platform, and with 6-drum and 8-drum low-profile platforms

Compliance

Meet U.S. EPA requirements for Secondary Spill Containment of Hazardous Materials 40CFR264.175.

Specifications

Certifications/Compliance EPA, 40 CFR 264.175, SPCC
Color Yellow
Capacity (English) 66 gal.
Capacity (Metric) 250 L
Material HDPE
Type 2 Drum Pallet, with Drain
For Use With (Application) Contain spills and overflows of hazardous materials, keeping floors contamination
Load Bearing Capacity (English) 4000 lb.
Load Bearing Capacity (Metric) 1814 kg
Shape Square
Width (English) 51 in.
Width (Metric) 130 cm
Height (English) 13.75 in.
Height (Metric) 35 cm
Depth (English) 26.25 in.
Depth (Metric) 67 cm
Weight (English) 52 lb.
Weight (Metric) 24 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.