Learn More
Eagle™ Drum Pallet Tarps
Catalog No. 18005 Shop All Eagle Manufacturing Products
Change view
Click to view available options
For Use With (Equipment):
2-drum spill control pallet
4-drum spill control pallet
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
18-005 4-drum spill control pallet
18-004 2-drum spill control pallet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-005 Supplier Eagle™ Supplier No. 1644
Prevent water build-up inside drum pallets
  • Protects drums
  • Weatherproof
  • For use with Eagle Two-Drum and Four-Drum Spill-Control Pallets
Specifications
Type Drum Pallet Tarp
For Use With (Application) Prevents water build up inside pallet and protects drums
For Use With (Equipment) 4-drum spill control pallet
Weight (English) 6 lb.
Weight (Metric) 2.7 kg