Learn More
MSC EAGLE DRUM TRAYS
Catalog No. 5000688645
Encompass
This drum tray dolly is designed to provide easy mobility for drum trays, facilitating the transportation of drums within a facility.
  • Load capacity of 1000 lb
  • Accommodates 30 gal to 55 gal drums
  • Constructed from steel with a powder coated frame
  • Equipped with four 3 in hard rubber swivel casters, some with side pedal brakes
  • Features an overall diameter of 31 in and a height of 7 in
Catalog No. 50-006-88645 Supplier MSC Supplier No. 99795163
This drum tray dolly is designed to provide easy mobility for drum trays, facilitating the transportation of drums within a facility.
  • Load capacity of 1000 lb
  • Accommodates 30 gal to 55 gal drums
  • Constructed from steel with a powder coated frame
  • Equipped with four 3 in hard rubber swivel casters, some with side pedal brakes
  • Features an overall diameter of 31 in and a height of 7 in