Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eagle™ Overpack Dolly
GREENER_CHOICE

Catalog No. 18869 Shop All Eagle Manufacturing Products
Catalog No. 18-869 Supplier Eagle™ Supplier No. 1698
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Safely moves salvage drums

  • Heavy-duty dolly holds 85 gal. (323L) and 95 gal. (361L) overpack salvage drums
  • Welded construction with rubber wheels

Order Info

Capacity: 1000 lb. (453.5kg). Dimensions: 27.5 x 7 in. (69.9 x 17.8cm). Weight: 30 lb. (13.6kg).

Specifications

Height (English) 7 in.
Product Type Drum Dolly
Color Black
Height (Metric) 17.7 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.