Learn More
MSC Eagle Safety Can, Diesel, 2 gal, Flame Arrester, 1 ea
Catalog No. 5000803011
Encompass
This Type I steel safety can for diesel is a rugged 2-gallon container designed to prevent fires and explosions by controlling flammable vapors. It provides a safe way to carry, pour, and store flammable liquids, color-coded yellow for clear communication of contents.
  • Seamless 24-gauge, hot-dip galvanized steel construction eliminates leaks and odor escape
  • Self-closing, leak-tested lid prevents spills
  • Double interlock, no-weld bottom seam safeguards against spills
  • Nitrile gasket automatically releases vapors for pressure relief
  • Non-sparking flame arrester prevents ignition
  • Comfort grip and trigger release handle for smooth pouring
  • Fixed steel handle for easy carrying
  • Trilingual safety label
Catalog No. 50-008-03011 Supplier MSC Supplier No. 86810736
This Type I steel safety can for diesel is a rugged 2-gallon container designed to prevent fires and explosions by controlling flammable vapors. It provides a safe way to carry, pour, and store flammable liquids, color-coded yellow for clear communication of contents.
  • Seamless 24-gauge, hot-dip galvanized steel construction eliminates leaks and odor escape
  • Self-closing, leak-tested lid prevents spills
  • Double interlock, no-weld bottom seam safeguards against spills
  • Nitrile gasket automatically releases vapors for pressure relief
  • Non-sparking flame arrester prevents ignition
  • Comfort grip and trigger release handle for smooth pouring
  • Fixed steel handle for easy carrying
  • Trilingual safety label