Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eagle™ Spill-Control Drum Funnel Cover
GREENER_CHOICE

Catalog No. 18884B Shop All Eagle Manufacturing Products
Change view
Click to view available options
For Use With (Equipment):
55 and 30 gal. Tight-head Drums, 5 gal. Pails
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
18-884B 55 and 30 gal. Tight-head Drums, 5 gal. Pails
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-884B Supplier Eagle™ Supplier No. 1664
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with Eagle Spill-Control Drum Funnel (see cat. no. 18600)

Compliance

Supports compliance with U.S. EPA 40CFR265.173.

Specifications

Certifications/Compliance U.S. EPA 40CFR265.173
Product Type Funnel Cover
Color Yellow
For Use With (Application) Spill Control
For Use With (Equipment) 55 and 30 gal. Tight-head Drums, 5 gal. Pails
Material HDPE
Quantity Ea.
Height (Metric) 177.8 mm
Diameter (Metric) Opening 460 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.