Learn More
Sigma Aldrich Fine Chemicals Biosciences Dulbecco's Modified Eagle's Medium/Nutrient Mixture F-12 Ham With L-glutamine, 15 mM HEPES, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture | MFCD00217417 | 6X500ML
Catalog No. NC1907008
Encompass
Dulbecco's Modified Eagle's Medium/Nutrient Mixture F-12 Ham With L-glutamine, 15 mM HEPES, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture | MFCD00217417 | 6X500ML
Catalog No. NC1907008 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D84376X500ML
Dulbecco's Modified Eagle's Medium/Nutrient Mixture F-12 Ham With L-glutamine, 15 mM HEPES, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture | MFCD00217417 | 6X500ML