Learn More
ATCC Eagle's Minimum Essential Medium (EMEM)
Catalog No. 502382632
Encompass
Eagle's minimum essential medium modified to contain Earle's Balanced Salt Solution, non-essential amino acids, 2 mM L-glutamine, 1 mM sodium pyruvate, and 1500 mg/L sodium bicarbonate. It has applications for cell culture, cell growth, and viability.
Catalog No. 50-238-2632 Supplier ATCC Supplier No. 302003
 Restricted Item
Add to Cart
Add to Cart
Eagle's minimum essential medium modified to contain Earle's Balanced Salt Solution, non-essential amino acids, 2 mM L-glutamine, 1 mM sodium pyruvate, and 1500 mg/L sodium bicarbonate. It has applications for cell culture, cell growth, and viability.