Learn More
Bioworld EAH SEPAROPOREAGAROSE) 4B
Catalog No. 50197672
Encompass
This item has a minimum order quantity of 2 per supplier requirements. BioWorld's EAH-Separopore was prepared by a covalent coupling of 1,6-diamino hexane to Separopore 4B via epoxy activation. The free terminal amino group allows for coupling of carboxyl-group containing ligands through amide bond formatio...
Catalog No. 50-197-672 Supplier Bioworld Supplier No. 201810071
This item has a minimum order quantity of 2 per supplier requirements. BioWorld's EAH-Separopore was prepared by a covalent coupling of 1,6-diamino hexane to Separopore 4B via epoxy activation. The free terminal amino group allows for coupling of carboxyl-group containing ligands through amide bond formatio...