Learn More
Bioworld EAH SEPAROPOREAGAROSE) 4B
Supplier:  Bioworld 201810073
Encompass
BioWorld's EAH-Separopore was prepared by a covalent coupling of 1,6-diamino hexane to Separopore 4B via epoxy activation. The free terminal amino group allows for coupling of carboxyl-group containing ligands through amide bond formatio...
Catalog No. 50-197-674