Learn More
Medchemexpress LLC EAI001 10mg | CAS No. : 892772-75-7
SDP
Catalog No. NC2949431
Encompass_Preferred
EAI001 is a potent, selective mutant epidermal growth factor receptor (EGFR) allosteric inhibitor with an IC50 value of 24 nM for EGFRL858R/T790M. EAI001 can be used for research of cancer.
Catalog No. NC2949431 Supplier Medchemexpress LLC Supplier No. HY10021410MG
EAI001 is a potent, selective mutant epidermal growth factor receptor (EGFR) allosteric inhibitor with an IC50 value of 24 nM for EGFRL858R/T790M. EAI001 can be used for research of cancer.