Learn More
Aviva Systems Biology Corp EAP30 Antibody - N-terminal region (ARP31531_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31531_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SNF8. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human EAP30. Alternate names: Dot3, EAP30, VPS22ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54256