Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Ear Plug Tray

Catalog No. 19129481 Shop All Brady Corporation Products
Change view
Click to view available options
Depth (English):
10.6 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Depth (English)
19-129-481 10.6 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-129-481 Supplier Brady™ Supplier No. PD228E
  • $176.00
    $157.65 / Each of 1

    Save $18.35

    10% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acrylic dispensers serve as a visual reminder for employees to wear their ear plugs. Conveniently mount near hazardous work areas to encourage compliance.

  • Clear acrylic ear plug dispenser tray holds a standard box of ear plugs
  • Front section catches spills and protects the corrugated dispenser
  • Use on table top or wall mount
  • Tray size is 10-5/8 H x 10-5/8 W x 10-5/8 D in.
  • Ear plugs not included

Specifications

Depth (English) 10.6 in.
Depth (Metric) 26.9 cm
For Use With (Application) Ear Plug Dispenser to be Wall Mounted or Placed on a Table Top Near Hazardous Work Areas.
Height (English) 10.6 in.
Height (Metric) 26.9 cm
Holds Ear plugs
Width (Metric) 26.9 cm
Width (English) 10.6 in.
Product Type Tray/Box Dispenser
Disposable Reusable
Material Acrylic
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.