Learn More
Med Vet International Ear Plugs 3M E-A-R Classic Cordless Large Yellow, 310-1101, 200 Pairs
Catalog No. 502829756
Encompass_Preferred
Reusable Pillow Pack helps keep earplugs clean when they are not being worn Classic™ Plus earplugs are slightly longer than regular E-A-R™ Classic™ earplugs to fit deeper into the ear canal and provide higher noise reduction. Flame-resistant and moisture-resistant Test compatible with 3M™ E-A-Rfit™ Validation System Noise reduction rating (NRR)* 33 dB. CSA Class AL
Catalog No. 50-282-9756 Supplier Med Vet International Supplier No. 775139BX
Reusable Pillow Pack helps keep earplugs clean when they are not being worn Classic™ Plus earplugs are slightly longer than regular E-A-R™ Classic™ earplugs to fit deeper into the ear canal and provide higher noise reduction. Flame-resistant and moisture-resistant Test compatible with 3M™ E-A-Rfit™ Validation System Noise reduction rating (NRR)* 33 dB. CSA Class AL