Learn More
Grainger Ear Plugs Corded Contoured-T 30dB PK5
Catalog No. 502765288
Encompass_Preferred
The 3M E-A-Rsoft Corded Contoured-T Earplugs, 30dB NRR, 5 Pair per Pack, 16R199 provide hearing protection with a Noise Reduction Rating (NRR) of 30 decibels. They feature a contoured T-shape that fits comfortably inside the ear canal. The corded design connects the earplugs to help prevent loss or misplacement. They are made of soft, energy-absorbing foam that is slow-recovery for comfort. The earplugs are designed for reuse and easy cleaning. They come 5 pairs per pack and are available in a standard size that fits most users. The 3M earplugs help block hazardous noise to protect hearing in loud work environments.
Catalog No. 50-276-5288 Supplier Grainger Supplier No. 16R199
The 3M E-A-Rsoft Corded Contoured-T Earplugs, 30dB NRR, 5 Pair per Pack, 16R199 provide hearing protection with a Noise Reduction Rating (NRR) of 30 decibels. They feature a contoured T-shape that fits comfortably inside the ear canal. The corded design connects the earplugs to help prevent loss or misplacement. They are made of soft, energy-absorbing foam that is slow-recovery for comfort. The earplugs are designed for reuse and easy cleaning. They come 5 pairs per pack and are available in a standard size that fits most users. The 3M earplugs help block hazardous noise to protect hearing in loud work environments.