Learn More
Grainger Ear Plugs Uncorded Contoured-T 32dB PK5
Catalog No. 502767287
Encompass_Preferred
The Grainger 16R196 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is thermally protected and rated for continuous duty applications. It has a C-face endshield for easy mounting and its enclosure helps protect against dirt and water. The 16R196 would be a good general purpose motor for light industrial, commercial, and residential applications.
Catalog No. 50-276-7287 Supplier Grainger Supplier No. 16R196
The Grainger 16R196 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is thermally protected and rated for continuous duty applications. It has a C-face endshield for easy mounting and its enclosure helps protect against dirt and water. The 16R196 would be a good general purpose motor for light industrial, commercial, and residential applications.