Learn More
Electron Microscopy Sciences Ear Punch, Stainless Steel, 1 each
SDP
Catalog No. 5032227
Encompass_Preferred
Made from stainless steel. Allows for you to make a 2 mm hole through the animal ear in just one second. It also can be used as a specimen puncher.
Catalog No. 50-322-27 Supplier Electron Microscopy Sciences Supplier No. 7289301
Made from stainless steel. Allows for you to make a 2 mm hole through the animal ear in just one second. It also can be used as a specimen puncher.