Learn More
Braintree Scientific Ear Punch Pincher with vial holder, 2.75mm punch
SDP
Catalog No. 501954762
Encompass
Ear Puncher This puncher relieves the stress on the wrist during ID marking as it requires a very gentle pressure to cut a hole. The stainless steel puncher has a sloped edge which provides a cut rather than a punch, which minimizes animal discomfort. Now updated with Eppendorf holder. The little puck holds the lid of the Eppendorf tube, so the biopsy sample falls perfectly into the 1,5 mL tube. For your convenience we also sell these tubes in set of 100pcs. The ear puncher is autoclavable but should be lubricated afterwards for a long and trouble-free usage. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4762 Supplier Braintree Scientific Supplier No. EPAT7075
Ear Puncher This puncher relieves the stress on the wrist during ID marking as it requires a very gentle pressure to cut a hole. The stainless steel puncher has a sloped edge which provides a cut rather than a punch, which minimizes animal discomfort. Now updated with Eppendorf holder. The little puck holds the lid of the Eppendorf tube, so the biopsy sample falls perfectly into the 1,5 mL tube. For your convenience we also sell these tubes in set of 100pcs. The ear puncher is autoclavable but should be lubricated afterwards for a long and trouble-free usage. **Sold for research purposes only and not for human use**