Learn More
Fine Science Tools Ear Punch, Stainless Steel & Anodized Aluminum, 11.5 cm
SDP
Catalog No. NC0264349
Encompass
Product: Ear Punch - Stainless Steel & Anodized Aluminum/2mm Punch Diameter/ Punch Diameter: 2mm/ Alloy: Stainless Steel & Anodized Aluminum/ Length: 11.5cm
Catalog No. NC0264349 Supplier Fine Science Tools Supplier No. 2421402
Product: Ear Punch - Stainless Steel & Anodized Aluminum/2mm Punch Diameter/ Punch Diameter: 2mm/ Alloy: Stainless Steel & Anodized Aluminum/ Length: 11.5cm