Learn More
Fine Science Tools EAR PUNCHER SS 2MM PUNCH DIA
SDP
Supplier:  Fine Science Tools 2491602
Encompass
Punch Diameter:2mmAlloy / Material: Stainless SteelLength:14cmThe Ear Puncher relieves stress on the wrist during ID marking as it requires only a very gentle pressure to cut a hole. The stainless steel puncher has a sloped edge that provides a cut rather than a punch, which minimizes animal discomfort.
Catalog No. NC1899423