Learn More
Med Vet International AbsorbENT Ear Wick, 9mm x 15mm, J1084, 50-Pack
Supplier: Med Vet International JOR1084
Ear wicks have been around a long time in human medicine. Recent improvements have made them a valuable adjunct to treating veterinary otitis externa. AbsorbENT sponges consist of interconnected open cells that are extremely soft and pliable. The interconnected cell structure produces an extremely strong packing that is highly absorbent and virtually lint and fiber free. AbsorbENT is designed as an aid in the treatment of otitis externa. When liquid medication is applied to the sterile, compressed sponge, it gently and evenly expands to fill the ear canal dispersing the medication and applying gentle pressure to the tissue. Expands rapidly for effective distribution of liquid medication Soft, pliable sponge enhances patient comfort while applying gentle pressure Compressed and sterile for rapid patient treatment Suture can be placed around the end for easy retrieval Suggested use: Insert Ear Wick and infuse appropriate antibiotic or corticosteroid into sponge.