Learn More
New England Biolabs, Inc. EarI – 2500 units
SDP
Catalog No. 50812187
Encompass
A restriction endonuclease that recognizes the sequence CTCTTCN^NNN_.
Catalog No. 50-812-187 Supplier New England Biolabs, Inc. Supplier No. R0528L
A restriction endonuclease that recognizes the sequence CTCTTCN^NNN_.